EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Human mitochondrial and cytosolic branched-chain aminotransferases are cysteine S-conjugate beta-lyases, but turnover leads to inactivation.
PMID 12504794
Abstract
The mitochondrial and cytosolic branched-chain aminotransferases (BCAT(m) and BCAT(c)) are homodimers in the fold type IV class of pyridoxal 5'-phosphate-containing enzymes that also contains D-amino acid aminotransferase and 4-amino-4-deoxychorismate lyase (a beta-lyase). Recombinant human BCAT(m) and BCAT(c) were shown to have beta-lyase activity toward three toxic cysteine S-conjugates [S-(1,1,2,2-tetrafluoroethyl)-L-cysteine, S-(1,2-dichlorovinyl)-L-cysteine, and S-(2-chloro-1,1,2-trifluoroethyl)-L-cysteine] and toward beta-chloro-L-alanine. Human BCAT(m) is a much more effective beta-chloro-L-alanine beta-lyase than two aminotransferases (cytosolic and mitochondrial isozymes of aspartate aminotransferase) previously shown to possess this activity. BCAT(m), but not BCAT(c), also exhibits measurable beta-lyase activity toward a relatively bulky cysteine S-conjugate [benzothiazolyl-L-cysteine]. Benzothiazolyl-L-cysteine, however, inhibits the L-leucine-alpha-ketoglutarate transamination reaction catalyzed by both enzymes. Inhibition was more pronounced with BCAT(m). In the presence of beta-lyase substrates and alpha-ketoisocaproate (the alpha-keto acid analogue of leucine), no transamination could be detected. Therefore, with an amino acid containing a good leaving group in the beta position, beta-elimination is greatly preferred over transamination. Both BCAT isozymes are rapidly inactivated by the beta-lyase substrates. The ratio of turnover to inactivation per monomer in the presence of toxic halogenated cysteine S-conjugates is approximately 170-280 for BCAT(m) and approximately 40-50 for BCAT(c). Mitochondrial enzymes of energy metabolism are especially vulnerable to thioacylation and inactivation by the reactive fragment released from toxic, halogenated cysteine S-conjugates such as S-(1,1,2,2-tetrafluoroethyl)-L-cysteine. The present results suggest that BCAT isozymes may contribute to the mitochondrial toxicity of these compounds by providing thioacylating fragments, but inactivation of the BCAT isozymes might also block essential metabolic pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4284
β-Chloro-D-alanine hydrochloride
C3H6ClNO2 · HCl
C9033
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
C3H6ClNO2 · HCl