EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Cannabidiol: an overview of some chemical and pharmacological aspects. Part I: chemical aspects.
PMID 12505688
Abstract
Over the last few years considerable attention has focused on cannabidiol (CBD), a major non-psychotropic constituent of Cannabis. In Part I of this review we present a condensed survey of the chemistry of CBD; in Part II, to be published later, we shall discuss the anti-convulsive, anti-anxiety, anti-psychotic, anti-nausea and anti-rheumatoid arthritic properties of CBD. CBD does not bind to the known cannabinoid receptors and its mechanism of action is yet unknown. In Part II we shall also present evidence that it is conceivable that, in part at least, its effects are due to its recently discovered inhibition of anandamide uptake and hydrolysis and to its anti-oxidative effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161977
N-Cyclohexylisopropylamine, 98%
C9H19N