EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Comparison of multiple DNA adduct types in tumor adjacent human lung tissue: effect of cigarette smoking.
PMID 12507931
Abstract
Cigarette smokers inhale a broad range of carcinogens derived from tobacco and its pyrolysis products, including free radicals, which induce oxidative stress and subsequent lipid peroxidation (LPO). Miscoding carcinogen-DNA adducts are formed by cigarette smoke constituents and are thought to initiate lung carcinogenesis. The presence of various types of DNA damage was therefore analyzed in tumor adjacent uninvolved lung tissues of 13 smoking and 11 non-smoking operated lung cancer patients. O(4)-ethylthymidine (O(4)etT), 1,N(6)-ethenodeoxyadenosine ( epsilon dA) and 3,N(4)-ethenodeoxycytidine ( epsilon dC) were determined by immuno-enriched (32)P-postlabeling. Polycyclic aromatic hydrocarbon (PAH)-DNA adducts were measured as diagonal radioactive zones after nuclease P1 enriched (32)P-postlabeling. Mean O(4)etT and PAH-DNA adduct levels were higher in lung DNA of smokers than of non-smokers (O(4)etT/10(8) thymidine: 3.8 versus 1.6, P < 0.01; PAH-DNA adducts/10(8) nucleotides: 11.2 versus 2.2, P < 0.01). Pulmonary etheno-DNA adduct levels did not differ between smokers and non-smokers, but large inter-individual variations were observed (80- and 250-fold differences for epsilon dA and epsilon dC, respectively). As all smokers (except one) refrained from smoking at least for 1 week before surgery, our results demonstrate the persistence of O(4)etT and PAH-DNA adducts in human lung. A positive correlation obtained between O(4)etT and PAH-DNA adducts (R = 0.65, P < 0.01) suggests that both adducts are formed from cigarette smoke as the main exposure source. We conclude that in addition to the DNA adducts derived from PAH and tobacco-specific nitrosamines, miscoding O(4)etT lesions are formed by cigarette smoke that contribute to the increased genomic instability and increased lung cancer risk in smokers.