EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A chromoreactand for the selective detection of HSO3- based on the reversible bisulfite addition reaction in polymer membranes.
PMID 12510279
Abstract
A new chromoreactand for the reversible optical detection of bisulfite anion is presented with a sensitivity in the range from 0.1 to 10 mM while no absorbance changes are observed for anions such as sulfate, chloride, phosphate or hydroxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
95647
Bisulfite ionophore, Selectophore
C29H42N4O3