EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical weed control of medicinal plant Bupleurum falcatum L].
PMID 12512434
Abstract
To select low-residue herbicide for cultivation of Bupleurum falcatum. Probing the effect of various kinds of herbicide on the budding, growth and yield of B. falcatum both in laboratory and in the fields. Haloxyfop acts slightly on the growth of B. falcarum, but effectively kills weeds of many kinds. Butralin is a good herbicide for Gramineae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36528
Butralin, PESTANAL®, analytical standard
C14H21N3O4
45817
Haloxyfop, PESTANAL®, analytical standard
C15H11ClF3NO4