EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Enantioselective oxidation of di-tert-butyl disulfide with a vanadium catalyst: progress toward mechanism elucidation.
PMID 12515473
Abstract
The mechanism of the oxidation of di-tert-butyl disulfide (1) to the chiral thiosulfinate (2) by H(2)O(2) catalyzed by bis(acetylacetonato)oxovanadium and a chiral Schiff-base ligand (3) has been investigated. Techniques included (51)V NMR spectroscopy, solvent effects on reaction enantioselectivity, and the isolation and full characterization of a 2:1 ligand-to-vanadium catalyst precursor. A model for the dramatic solvent effect on the enantioselectivity of this reaction was developed, based on the identification of a competing nonselective oxidation pathway. From this model, strategies for limiting this competing pathway were developed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247545
Di-tert-butyl disulfide, 97%
C8H18S2