EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, Thrombosis, and Vascular Biology
Effects of chronic treatment with L-arginine on atherosclerosis in apoE knockout and apoE/inducible NO synthase double-knockout mice.
PMID 12524231