EMAIL THIS PAGE TO A FRIEND
Fish & shellfish immunology
The promoter of the classical MHC class I locus in rainbow trout (Oncorhynchus mykiss).
PMID 12526881
Abstract
In rainbow trout only a single classical major histocompatibility complex (MHC) class I locus has been identified. In previous studies it was shown that this locus, Onmy-UBA, is characterised by extensive polymorphism and ubiquitous expression. In the present study the sequence of the promoter region of Onmy-UBA was analysed. Within this region several fragments for transcription regulation could be detected, including interferon-sensitive enhancer elements. The promoter region is very similar to that of classical MHC class I in higher vertebrates. In agreement with the interferon sensitive elements identified in its promoter, Onmy-UBA expression in rainbow trout gonad cells (RTG-2 cell line) was up-regulated after infection with infectious haematopoietic necrosis virus (IHNV).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
RTG-2, 90102529