EMAIL THIS PAGE TO A FRIEND
Life sciences
Anticlastogenic effect of Spirulina maxima extract on the micronuclei induced by maleic hydrazide in Tradescantia.
PMID 12527032
Abstract
The aim of this investigation was to determine if extracts of Spirulina maxima reduce the genotoxic damage induced by maleic hydrazide (MH) using the Tradescantia biosssay. Two types of extracts from the alga were prepared: an aqueous extract with two different concentrations, 100 and 500 mg/ml, and a second one, the extract of a 1% solution of dimethyl sulfoxide (DMSO) which corresponded to 100 mg/ml of the alga. The capacity of MH to induce micronuclei (MN) was initially established by administering 0.005, 0.01, and 0.015 mg/ml of the chemical to the Tradescantia inflorescences, and observing its effect after 24 h.The results of this experiment showed a significant MN increase with the two high concentrations tested, although no dose-response effect was observed. For the anticlastogenic assay, the extracts of Spirulina were applied to the inflorescences alone or immediately before the application of MH (0.01 mg/ml) and the induced MN were observed 24 h later. We found that none of the extracts increased the MN level with respect to the untreated plants; also, that MH more or less doubled the basal micronuclei frequency, and finally, that all tested extracts reduced the genotoxic damage caused by MH. The inhibitory indices obtained for the aqueous extracts (100 and 500 mg/ml) and for the DMSO extract were respectively 59, 85, and 56.3%. These data indicate that Spirulina is an anticlastogenic agent and suggest that it is advisable to extend studies on this matter using other biological models.
Related Materials