EMAIL THIS PAGE TO A FRIEND
FEBS letters
Isolation of a new dual-functional caffeine synthase gene encoding an enzyme for the conversion of 7-methylxanthine to caffeine from coffee (Coffea arabica L.).
PMID 12527364
Abstract
In coffee and tea plants, caffeine is synthesized from xanthosine via a pathway that includes three methylation steps. We report the isolation of a bifunctional coffee caffeine synthase (CCS1) clone from coffee endosperm by reverse transcription-polymerase chain reaction (RT-PCR) and rapid amplification of cDNA ends (RACE) technique using previously reported sequence information for theobromine synthases (CTSs). The predicted amino acid sequences of CCS1 are more than 80% identical to CTSs and are about 40% similar to those of tea caffeine synthase (TCS1). Interestingly, CCS1 has dual methylation activity like tea TCS1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69723
7-Methylxanthine, ≥98.0% (HPLC)
C6H6N4O2