EMAIL THIS PAGE TO A FRIEND
Sheng li ke xue jin zhan [Progress in physiology]
[Relationship between oxysterols and atherosclerosis].
PMID 12532844
Abstract
Oxysterols, being oxygenated derivatives of cholesterol, are as many kinds as one hundred. They are found in the foodstuffs, the blood and arterial tissues of animals with hypercholesterolemia, the human atheroma, the foam cell from atherosclerotic tissues, as well as in the oxidized low density lipoprotein. The results of experiments have demonstrated that oxysterols are cytotoxic and they are injurious to a variety of cells including vascular endothelial cells, smooth muscle cells and monocyte/macrophages, suggesting that oxysterols could play a key role in the initiation and pathogenesis of atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2773
Cholesterol 5α,6α-epoxide
C27H46O2
C2648
Cholesterol 5β,6β-epoxide, ≥98%
C27H46O2