EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of the butyl 2,4-dichlorophenoxyacetate in the workshop by gas chromatography].
PMID 12541598
Abstract
The new method for determining the content of 2,4-dichlorophenoxyacetic acid butyl ester in the workshop by using gas chromatography is presented. The experiments for precision and efficiency have been performed. The range of determination is 0.05-2.5 mg/m3. The minimum detectable mass concentration is 25 micrograms/m3. The method meets the requirement of the "monitoring and researching standard for toxic substances", and shows that it can be applied in routine analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45732
2,4-D 1-butyl ester, PESTANAL®, analytical standard
C12H14Cl2O3