EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Synthesis of a new cyclodextrin derivative and its application as CGC chiral stationary phase in determination of enantiomeric excess].
PMID 12541614
Abstract
A new cyclodextrin (CD) derivative, 2,6-di-O-benzyl-3-O-valeryl-beta-CD, was synthesized and characterized by 1H NMR and IR. Using the beta-CD derivative as chiral stationary phase of capillary gas chromatography, one chiral column was prepared. On this column, the enantiomeric excesses (e.e.) of 1-(2,4-dichlorophenyl) ethanol obtained by asymmetric catalytic hydrogenation of 2,4-dichloroacetophone and trans-3-propyloxiranemethanol synthesized by Sharpless epoxidation of trans-2-hexenol were determined and the catalytic reactions were evaluated. The results are satisfactory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178373
2′,4′-Dichloroacetophenone, 96%
C8H6Cl2O