EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Study on the analysis of alpha-naphthylacetic acid].
PMID 12541752
Abstract
This paper introduces a method for the determination of alpha-naphthylacetic acid with HPLC. Under the reversed-phase condition, puting ion-pair reagent IPR-A in the mobile phase of methanol-water, an unknown peak was separated. The recovery was 100.2% and the RSD was 0.30%. In comparing with conventional HPLC using methanol-water as mobile phase and GC methods, the results were more dependable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8505
1-Naphthyl acetate, ≥98% (C)
C12H10O2