EMAIL THIS PAGE TO A FRIEND
Journal of biochemical and biophysical methods
Separation of oxazepam, lorazepam, and temazepam enantiomers by HPLC on a derivatized cyclodextrin-bonded phase: application to the determination of oxazepam in plasma.
PMID 12543505
Abstract
The enantioselective high-performance liquid chromatography (HPLC) of three racemic 3-hydroxybenzodiazepines, oxazepam (Oxa), lorazepam (Lor), and temazepam (Tem), is a difficult operation because of the spontaneous chiral inversion in polar solvent. To solve this problem, we have developed an HPLC method based on a chiral Cyclobond I-2000 RSP column, maintained at 12 degrees C, and a reversed mobile phase (acetonitrile in 1% triethylamine acetate buffer, TEAA) at a flow rate of 0.4 ml/min. Peaks were detected by a photodiode-array detector at 230 nm for quantification and by an optical rotation detector for identification of (+) and (-) enantiomers. The results showed that peak resolutions of Oxa, Lor, and Tem enantiomers, analyzed under the same conditions, were 3.2, 2.0, and 1.8, respectively. For the determination of Oxa enantiomers in plasma of rabbits, extraction with diethyl ether at pH 1.5, a polar organic mobile phase, and a Cyclobond I-2000 SP column were used. Other analytical conditions were the same as previously described. Blood samples were immediately cooled at 4 degrees C and centrifuged at 0 degrees C for the collection of plasma. The results showed a difference in plasma S(+)- and R(-)-oxazepam concentrations in rabbits. No racemization of S(+)- or R(-)-Oxa enantiomers, added alone to blank plasma, was observed after extraction and enantioselective HPLC analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20018AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
20019AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
20020AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
20022AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
20024AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
20034AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
22024AST Astec® CYCLOBOND® I 2000 Chiral HPLC Column, 10 μm particle size, L × I.D. 25 cm × 4.6 mm
21100AST Astec® CYCLOBOND® I 2000 Chiral HPLC Guard, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
20318AST Astec® CYCLOBOND® I 2000 RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
20324AST Astec® CYCLOBOND® I 2000 RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
20334AST Astec® CYCLOBOND® I 2000 RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
21013AST Astec® CYCLOBOND® I 2000 RSP Chiral HPLC Guard, Column, 5 μm particle size, L × I.D. 2 cm × 1 mm
21103AST Astec® CYCLOBOND® I 2000 RSP Chiral HPLC Guard, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
20224AST Astec® CYCLOBOND® I 2000 SP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
21102AST Astec® CYCLOBOND® I 2000 SP Chiral HPLC Guard, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm