EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Screening of interleukin-2 production inhibitor with mouse thymoma EL4 cells.
PMID 12546423
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4368
Piericidin A from microbial source, >95% (HPLC), DMSO solution
C25H37NO4