EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Study on qualitative analysis method of high performance liquid chromatography by simultaneous multi-wavelength detection].
PMID 12549162
Abstract
In this paper, we introduce an HPLC qualitative method, that is, multiwavelength detection qualitative analysis method. According to Beer's law, for a certain compounds the ratio of peak areas at different wavelengths is a constant. Comparing the ratio of peak areas of a component at different wavelengths with that of the standard by means of simultaneous multi-wavelength detection, we established a convenient qualitative method. This method has been applied successfully to qualitative analysis of 18 aromatic amines in the prohibited azo dyes.