EMAIL THIS PAGE TO A FRIEND
Wei sheng wu xue bao = Acta microbiologica Sinica
[Enzymatic synthesis of cephradine].
PMID 12549419
Abstract
Cephradine was synthesized by gamma-alumina-immobilized form of the penicillin G acylase of Bacillus megaterium with D-phenyglycine methylester hydrochloride (CH DGME.HCl) as acyl donor and 7-aminodeacetoxycephalosporanic acid (7-ADCA) as acyl acceptor. 0.1 g of 7-ADCA was dissolved by adding 2.5 ml of distilled water and about 0.25 ml of 2 mol/L NaOH in a 25 ml flask. To the solution, after 0.25 g of CHDGME.HCl was added, 0.1 mol/L phosphate-0.05 mol/L citric acid buffer, pH 7.5 was added to result in a volume of 5 ml with pH 7.5 Then 1 g(220 IU) of immobilized enzyme was added. The flask was shaken on a rotary shaker at 110 r/min and 25 degrees C for 5 h. The conversion rate of 7-ADCA was 81%. In an expanded experiment in 500 ml of reactive volume, 11.8 g of cephradine was obtained from 10 g of 7-ADCA. The conversion rate of 7-ADCA was 80% with about 87% yield of cephradine. Enzymatic synthesis was inhibited in varying degrees by phenylacetic acid, phenoxyacetic acid and cephalosporin G.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8398
7-Aminodesacetoxycephalosporanic acid
C8H10N2O3S