EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Two Hsp70 family members expressed in atherosclerotic lesions.
PMID 12552099
Abstract
Gene expression profiling was carried out comparing Con A elicited peritoneal macrophages from C57BL6 and FVBN wild-type and apolipoprotein (apo)E knockout mice. An EST, was expressed at higher levels in C57BL6 compared with FVBN mice. mapped to an atherosclerosis susceptibility locus on chromosome 19 revealed in an intercross between atherosclerosis-susceptible C57BL6 and atherosclerosis-resistant FVBN apoE knockout mice. A combination of database search and Northern analysis confirmed that corresponded to 3'-UTR of a hitherto predicted gene, named HspA12A. Blasting the National Center for Biotechnology Information database revealed a closely related homologue, HspA12B. HspA12A and -B have very close human homologues. TaqMan analysis confirmed the increased HspA12A expression (2.6-fold) in elicited peritoneal macrophages from C57BL6 compared with FVBN mice. TaqMan analysis also revealed increased HspA12A and HspA12B expression (87- and 6-fold, respectively) in lesional versus nonlesional portions of the thoracic aorta from C57BL6 apoE knockout mice on a chow diet. In situ hybridization confirmed that both genes were expressed within lesions but not within nonlesional aortic tissue. Blasting of HspA12A and HspA12B against the National Center for Biotechnology Information database (NR) revealed a hit with the Conserved Domain database for Hsp70 (pfam00012.5, Hsp70). Both genes appear to contain an atypical Hsp70 ATPase domain. The BLAST search also revealed that both genes were more similar to primitive eukaryote and prokaryote than mammalian Hsp70s, making these two genes distant members of the mammalian Hsp70 family. In summary, we describe two genes that code for a subfamily of Hsp70 proteins that may be involved in atherosclerosis susceptibility.