EMAIL THIS PAGE TO A FRIEND
Indian pediatrics
Infantile cystinosis.
PMID 12554913
Abstract
Ten patients of nephropathic cystinosis were admitted during the period 1995-2000. Their mean age was 12 months. The signs of failure to thrive and advanced rickets were seen in all patients. Other features included polyuria, polydipsia, pathologic fractures and deafness. Laboratory findings included glucosuria, hyposthenuria, hypocalcemia, proteinuria and azotemia. Therapy with phosphocysteamine showed marked clinical improvement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8397
Cysteamine S-phosphate sodium salt, analytical standard
C2H7NNaO3PS