EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective palladium-catalyzed arylation of 3-carboalkoxy furan and thiophene.
PMID 12556177
Abstract
[reaction: see text] The regioselective palladium(0)-catalyzed arylation of 3-furoate and 3-thiophenecarboxylate esters with aryl bromides is described. Conditions were developed that allow for the selective synthesis of either 2-aryl or 5-aryl products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164542
Ethyl 3-furoate, 98%
C7H8O3