EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Modification of the philanthotoxin-343 polyamine moiety results in different structure-activity profiles at muscle nicotinic ACh, NMDA and AMPA receptors.
PMID 12559123
Abstract
Voltage-dependent, non-competitive inhibition by philanthotoxin-343 (PhTX-343) analogues, with reduced charge or length, of nicotinic acetylcholine receptors (nAChR) of TE671 cells and ionotropic glutamate receptors (N-methyl-D-aspartate receptors (NMDAR) and alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptors (AMPAR)) expressed in Xenopus oocytes from rat brain RNA was investigated. At nAChR, analogues with single amine-to-methylene or amine-to-ether substitutions had similar potencies to PhTX-343 (IC(50)=16.6 microM at -100 mV) whereas PhTX-(12), in which both secondary amino groups of PhTX-343 were replaced by methylenes, was more potent than PhTX-343 (IC(50)=0.93 microM at -100 mV). Truncated analogues of PhTX-343 were less potent. Inhibition by all analogues was voltage-dependent. PhTX-343 (IC(50)=2.01 microM at -80 mV) was the most potent inhibitor of NMDAR. At AMPAR, most analogues were equipotent with PhTX-343 (IC(50)=0.46 microM at -80 mV), apart from PhTX-83, which was more potent (IC(50)=0.032 microM at -80 mV), and PhTX-(12) and 4,9-dioxa-PhTX-(12), which were less potent (IC(50)s>300 microM at -80 mV). These studies show that PhTX-(12) is a selective nAChR inhibitor and PhTX-83 is a selective AMPAR antagonist.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P206
Philanthotoxin 343 tris(trifluoroacetate) salt, solid
C23H41N5O3 · 3C2HF3O2