EMAIL THIS PAGE TO A FRIEND
Uchu Seibutsu Kagaku
[Effect of simulated microgravity conditions on auxin polar transport of transgenic Arabidopsis].
PMID 12561874
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286281
Indole-3-acetamide, 98%
C10H10N2O