EMAIL THIS PAGE TO A FRIEND
Zhongguo ji sheng chong xue yu ji sheng chong bing za zhi = Chinese journal of parasitology & parasitic diseases
[Mollusciciding action and toxicity of bromoacetamide].
PMID 12563778