EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Analysis of chemical constituents in the essential oil from Evodia fargesii by GC/MS].
PMID 12575060
Abstract
In the essential oil from Evodia fargesii Dode, 33 constituents were identified by means of GC-MS-DS. The content of these compounds are 91.0% in the essential oil of this plant. The main compounds are eighteen terpenoids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22135
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)
C15H26O
W521418
Cedrol, redistilled
C15H26O