EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Assay of three hydrolyzable tannins in Fructus Chebulae from different habitats by RP-HPLC].
PMID 12575084
Abstract
Three hydrolyzable tannins chebulinic acid (I), chebulagic acid(II) and 1,3, 6-tri-O-galloyl-beta-D-glucose (III) in Fructus Chebulae from different habitats were determined by RP-HPLC method. The contents of I and II were obviously interrelated with the variety and characteristics of Fructus Chebulae. It's suitable to use I and II as indexes in quality evaluation of the crude drug of Fructus Chebulae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69455
Chebulagic acid, analytical standard
C41H30O27
SMB00271 Chebulinic acid, ≥85% (LC/MS-ELSD)
C41H32O27