EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on chemical constituents of the fruits of Rosa bracteata var. bracteata].
PMID 12575158
Abstract
The chemical constituents of Rosa bracteata Wendl. var. bracteata were studied for the first time. Six compounds were isolated from the fruits of the plant, and four of them have been identified as 5-hydroxymethyl furfural, 5-acetoxymethyl furfural, beta-sitosterol, oleanolic acid. 5-hydroxymethy furfural and 5-acetoxymethyl furfural were obtained from genus Rosa L. for the first time. Otherwise, a white wax was obtained by silica gel column chromatography, from which four main constituents were identified as nonadecene, n-docosane, hentriacontane and hexatriacontane by GC-MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51529
Hentriacontane, analytical standard
C31H64