EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Study on effect of qingkailing injection and its active principle in inducing cell apoptosis in human acute promyelocytic leukemia].
PMID 12575380
Abstract
To investigate the mechanism of Qingkailing (QKL) Injection in treating acute promyelocytic leukemia. Using MTT technique, cell morphologic method, DNA gel electrophoresis and flow-cytometry to study the human acute promyelocytic leukemia (HL-60) cell apoptosis induced by QKL and its active principle. QKL and its active principle, Baicalin and hyodeoxycholic acid, showed strong cytotoxicity in inhibiting HL-60 cell, the Bezoar cholic acid showed a weaker effect. Apoptosis could be induced after being treated for 6 hrs by the former three principles, showing a typical apoptosis peak under flow-cytometry, but could not be induced by the latter one. QKL could induce leukemia cell apoptosis in vitro, which may be one of the mechanisms of QKLI in curing acute promyelocytic leukemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3878
Hyodeoxycholic acid, ≥98%
C24H40O4