EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Clinical and experimental study of shuangcao tuihuang granule-1 in treating severe jaundice of acute icterohepatitis].
PMID 12575547
Abstract
To observe the effect of Shuangcao Tuihuang Granule-1 (SCTH-1) in treating severe jaundice of acute icterohepatitis and to study its mechanism. Thirty-four patients with severe jaundice of acute icterohepatitis were treated with SCTH-1, their therapeutic effects were analyzed. In the experimental studies, the influences of SCTH-1 on acute liver injury, liver superoxide dismutase (SOD) and serum ALT and AST were monitored. The markedly effective rate and effective rate of the treated group were 73.5% and 23.5% respectively. The markedly effective rate of the treated group was obviously higher than that of the control group (P < 0.05). Experimental study revealed that SCTH-1 could reduce the level of serum transaminase and inflammation of liver tissue in mice with acute liver damage. In addition, SCTH-1 could raise the activity of liver SOD (P < 0.05). SCTH-1 could accelerate the jaundice subsidence, improve the liver function and ameliorate the liver injury, its mechanism was possibly correlated with raising SOD activity, scavenging the oxygen free radicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462233
D-Galactal, 95%
C6H10O4