EMAIL THIS PAGE TO A FRIEND
Nature immunology
Homozygous loss of ICOS is associated with adult-onset common variable immunodeficiency.
PMID 12577056
Abstract
No genetic defect is known to cause common variable immunodeficiency (CVID), a heterogeneous human disorder leading to adult-onset panhypogammaglobulinemia. In a search for CVID candidate proteins, we found four of 32 patients to lack ICOS, the "inducible costimulator" on activated T cells, due to an inherited homozygous deletion in the ICOS gene. T cells from these individuals were normal with regard to subset distribution, activation, cytokine production and proliferation. In contrast, naive, switched and memory B cells were reduced. The phenotype of human ICOS deficiency, which differs in key aspects from that of the ICOS-/- mouse, suggests a critical involvement of ICOS in T cell help for late B cell differentiation, class-switching and memory B cell generation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10433 CompoZr® Knockout ZFN Kit, Human ICOS (NM_012092)