EMAIL THIS PAGE TO A FRIEND
The Journal of Thoracic and Cardiovascular Surgery
Survival and death-promoting events after transient spinal cord ischemia in rabbits: induction of Akt and caspase3 in motor neurons.
PMID 12579107