EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Analysis of tannins in Fructus Chebulae and its confusion varieties by HPCE].
PMID 12580059
Abstract
To analyze the hydrolyzable tannins-chebulinic acid (I) and chebulagic acid(II) in Fructus Chebulae and its confusion varieties by using high performance capillary electrophoresis (HPCE) method. Using a capillary (375 microns OD x 50 microns ID; 81.5 cm x 61.5 cm) and a power supply set at 24 kV, with phosphate-borate buffer containing 20 mmol.L-1 Na2HPO4-60 mmol.L-1 boric acid and a UV detector at 280 nm, sample solution was loaded in decompression mode at the positive end of the capillary, the loading time was 5 s. The linear ranges of I and II were 0.0842-0.842 and 0.842 and 0.0940-0.940 mg.mL-1 respectively, the correlation coefficient were 0.9966 and 0.9957, the average recoveries were 95.6% (RSD = 4.0%, n = 5) and 95.0% (RSD = 4.4%, n = 5), the RSDs (n = 5) of measurement precision test were 2.2% and 1.7%, the RSDs (n = 6) of reproduction test were 5.4% and 4.0% respectively. The contents of I and II were obviously interrelated with the variety and characteristics of Fructus Chebulae, the contents of I and II in the confusion varieties of Fructus Chebulae were very low. It is suitable to use I and II as the criterion in quality evaluation of Fructus Chebulae, and the HPCE method is effective for quality evaluation of the crude Fructus Chebulae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69455
Chebulagic acid, analytical standard
C41H30O27
SMB00271 Chebulinic acid, ≥85% (LC/MS-ELSD)
C41H32O27