EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Mapping of chorismate mutase and prephenate dehydrogenase domains in the Escherichia coli T-protein.
PMID 12581215
Abstract
The Escherichia coli bifunctional T-protein transforms chorismic acid to p-hydroxyphenylpyruvic acid in the l-tyrosine biosynthetic pathway. The 373 amino acid T-protein is a homodimer that exhibits chorismate mutase (CM) and prephenate dehydrogenase (PDH) activities, both of which are feedback-inhibited by tyrosine. Fifteen genes coding for the T-protein and various fragments thereof were constructed and successfully expressed in order to characterize the CM, PDH and regulatory domains. Residues 1-88 constituted a functional CM domain, which was also dimeric. Both the PDH and the feedback-inhibition activities were localized in residues 94-373, but could not be separated into discrete domains. The activities of cloned CM and PDH domains were comparatively low, suggesting some cooperative interactions in the native state. Activity data further indicate that the PDH domain, in which NAD, prephenate and tyrosine binding sites were present, was more unstable than the CM domain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114286
4-Hydroxyphenylpyruvic acid, 98%
C9H8O4