EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Total synthesis of (s)-(+)-citreofuran by ring closing alkyne metathesis.
PMID 12585897
Abstract
A concise total synthesis of citreofuran 4 is described, a structurally unique octaketide derivative belonging to the curvularin family. Key steps involve the elaboration of orsellinic acid methyl ester 5 to acid 14, which converts, on attempted formation of the corresponding acid chloride, to the 3-alkoxyisocoumarin derivative 20. This heterocycle can be used as an activated ester to give ketone 21 on treatment with 3-pentynylmagnesium bromide in the presence of TMSCl as the activating agent. Ring- closing alkyne metathesis (RCAM) of diyne 21 catalyzed by (tBuO)(3)W[triple bond]CCMe(3) affords the strained cycloalkyne 22. Treatment with acid renders its triple bond susceptible to nucleophilic attack by the adjacent carbonyl group, thus leading to a transannular cycloaromatization with formation of the intact skeleton of citreofuran. An X-ray crystallographic study reveals conformational details about this natural product. Finally, it is shown that 4 as well as its protected precursor 23 are able to cleave double-stranded DNA under oxidative conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0772
Curvularin, from Penicillium citrinum, ≥98% (HPLC)
C16H20O5