EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Catalytic carbonylation of functionalized diamines: application to the core structure of DMP 323 and DMP 450.
PMID 12585919
Abstract
W(CO)(6)-catalyzed carbonylation provides an alternative to phosgene or phosgene derivatives such as 1,1-carbonyldiimidazole (CDI) for the conversion of amines to ureas. As an illustration, the core structure of the HIV protease inhibitors DMP 323 and DMP 450 has been prepared by catalytic carbonylation of diamine intermediates from the original syntheses.