EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Determination of residues of organic chloride pesticide in Chinese medicinal materials with capillary gas chromatography].
PMID 12587153
Abstract
Residues of organic chloride pesticide were separated and detected successfully with high performance capillary column and high sensitive electron capture detector, such as hexachloro-cyclohexane, dichloro-diphenyl-trichloroethane, pentachloronitrobenzene. The method can be used for the determination of residues of organic chloride pesticide in Chinese medicinal materials, and the residue enriching in the different part of plant body was tested preliminary.