EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Pharmacological comparison between virosecurinine and securinine].
PMID 12587161
Abstract
Pharmacological activities of virosecurinine (Vse) and securinine (Sec) were studied. The results showed that acute toxicity of Vse was 1/13.6 that of Sec, and Vse had no convulsive effects on rats or frogs, while Sec had. The results also showed that Vse and Sec could elevate blood pressure and excite respiration in cats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0055
Securinine, ≥98% (HPLC)
C13H15NO2