EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Visualization of identified GFP-expressing cells by light and electron microscopy.
PMID 12588954
Abstract
We have developed a procedure for visualizing GFP expression in fixed tissue after embedding in LR White. We find that GFP fluorescence survives fixation in 4% paraformaldehyde/0.1% glutaraldehyde and can be visualized directly by fluorescence microscopy in unstained, 1 microm sections of LR White-embedded material. The antigenicity of the GFP is retained in these preparations, so that GFP localization can be visualized in the electron microscope after immunogold labeling with anti-GFP antibodies. The ultrastructural morphology of tissue fixed and embedded by this protocol is of quality sufficient for subcellular localization of GFP. Thus, expression of GFP constructs can be visualized in living tissue and the same cells relocated in semithin sections. Furthermore, semithin sections can be used to locate GFP-expressing cells for examination by immunoelectron microscopy of the same material after thin sectioning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9774 LR white acrylic resins, Medium