EMAIL THIS PAGE TO A FRIEND
Gene
SF4 and SFRS14, two related putative splicing factors on human chromosome 19p13.11.
PMID 12594045
Abstract
The splicing of nascent mRNA precursors is an essential step for the expression of all intron-containing eukaryotic genes. Removal of intron sequences from nascent transcripts is mediated by the spliceosome, a large multicomponent complex. We describe here the identification of two genes encoding related, putative splicing factors on human chromosome 19p13.11, SF4 (splicing factor 4) and SFRS14 (splicing factor arginine/serine-rich 14). Both genes encode proteins containing a SURP motif; this domain is found in several splicing proteins including Drosophila alternative splicing regulator, suppressor-of-white-apricot (SWAP) and the yeast splicing factor, prp21p. In addition, SF4 and SFRS14 contain a G-patch domain at their C-termini, a motif present in a large number of eukaryotic RNA-binding proteins. SFRS14 also contains an N-terminal region that is rich in arginine/serine residues, suggesting SFRS14 is a novel member of the SR-related family of pre-mRNA processing factors. We have also identified the mouse orthologues of SF4 and SFRS14, based on conserved domain organization and high sequence similarity. Interestingly, SFRS14 undergoes alternative 3'-end processing events that are conserved between human and mouse, suggesting a functional significance.