EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Studies of the mechanism of action of the aerolysin-like hemolysin of Aeromonas sobria in stimulating T84 cells to produce cyclic AMP.
PMID 12595476
Abstract
We previously reported that the aerolysin-like hemolysin of Aeromonas sobria stimulates T84 cells to produce cyclic AMP, which then emerges in the culture medium. In order to clarify the mechanism of action of the hemolysin, we examined the involvement of adenosine nucleotide. The results show that the hemolysin stimulates T84 cells to release ATP, which is then converted to adenosine by ectonucleotidase. The adenosine generated might stimulate the P1 adenosine receptors of T84 cells to produce cyclic AMP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X103
Xanthine amine congener, ≥96% (HPLC)
C21H28N6O4