EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Identification of (3S, 9R)- and (3S, 9S)-megastigma-6,7-dien-3,5,9-triol 9-O-beta-D-glucopyranosides as damascenone progenitors in the flowers of Rosa damascena Mill.
PMID 12596869
Abstract
The progenitors of damascenone (1), the most intensive C13-norisoprenoid volatile aroma constituent of rose essential oil, were surveyed in the flowers of Rosa damascena Mill. Besides 9-O-beta-D-glucopyranosyl-3-hydroxy-7,8-didehydro-beta-ionol (4b), a stable progenitor already isolated from the residual water after steam distillation of flowers of R. damascena Mill., two labile progenitors were identified to be (3S, 9R)- and (3S, 9S)-megastigma-6,7-dien-3,5,9-triol 9-O-beta-D-glucopyranosides (2b) based on their synthesis and HPLC-MS analytical data. Compound 2b gave damascenone (1), 3-hydroxy-beta-damascone (3) and 4b upon heating under acidic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W342017
Damascenone, natural, 1.1-1.4 wt. % (190 proof ethanol), FG
C13H18O