EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Estrogen therapy--high-dose intravenous diethylstilbestrol diphosphate therapy for advanced or hormone refractory prostate cancer].
PMID 12599571