EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Nonradioactive methods for the assay of phosphoinositide 3-kinases and phosphoinositide phosphatases and selective detection of signaling lipids in cell and tissue extracts.
PMID 12605860
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14-558-M
PI3 Kinase (p120γ), 20 µg, For use in Kinase Assays.