EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Modeling of prebiotic catalysis with adenylated polymeric templates: crystal structure studies and kinetic characterization of template-assisted phosphate ester hydrolysis.
PMID 12613036
Abstract
We have synthesized and characterized novel, copper-metalated, polymeric templates that contain adenine nucleobases. These promote hydrolysis of non-natural and natural phosphate ester substrates in a highly efficient and catalytic fashion. The crystal structure of the cooper-containing adenylated monomer reveals the formation of a polymeric array, through coordination to both N1 and N7 atoms. Possible implications of these studies for prebiotic catalysis, involving synergism between adenine and copper ions, are also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9376
Adenosine 2′:3′-cyclic monophosphate sodium salt, ≥93%
C10H11N5NaO6P