EMAIL THIS PAGE TO A FRIEND
Dentistry today
Capping carious exposed pulps with potassium nitrate, dimethyl isosorbide, polycarboxylate cement.
PMID 12616888
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247898
Isosorbide dimethyl ether, 98%
C8H14O4