EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Oxidative biosynthesis of phenylbenzoisochromenones from phenylphenalenones.
PMID 12620342
Abstract
13C NMR analysis demonstrated incorporation of two 13C labelled phenylalanine units into phenylphenalenones and phenylbenzoisochromenones co-occurring in Wachendorfia thyrsiflora. These results suggest oxidative formation of phenylbenzoisochromenones following a late branching from a common phenylphenalenone biosynthetic pathway. A dioxygenase-type mechanism, followed by decarboxylation, is suggested for the key steps of this conversion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P10801
Perinaphthenone, 97%
C13H8O