EMAIL THIS PAGE TO A FRIEND
The British journal of nutrition
Folic acid treatment reduces elevated plasma levels of asymmetric dimethylarginine in hyperhomocysteinaemic subjects.
PMID 12628031
Abstract
Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of NO synthase, has been suggested to be a novel risk factor for endothelial dysfunction. It has previously been reported that hyperhomocysteinaemia may be associated with impaired endothelium-dependent vasodilation and reduced plasma level of NO-derived endproducts (NOx). In the present study, plasma levels of arginine and ADMA were measured in twenty-one healthy control subjects, and in twenty-one hyperhomocysteinaemic subjects before and after 6 weeks and 12 months of folic acid supplementation, and compared with previously measured plasma NOx values in the hyperhomocysteinaemic subjects. Compared with control subjects, hyperhomocysteinaemic subjects had higher plasma levels of arginine and ADMA. More importantly, folic acid therapy significantly reduced plasma levels of arginine and ADMA. Furthermore, plasma levels of arginine and ADMA were positively correlated with plasma homocysteine levels and negatively correlated with plasma folate, as well as negatively correlated with plasma NOx. Our results suggest that ADMA may be a mediator of the atherogenic effects of homocysteine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01769
Folic acid, tested according to Ph.Eur.
C19H19N7O6