EMAIL THIS PAGE TO A FRIEND
Journal of virology
Epstein-Barr virus regulates STAT1 through latent membrane protein 1.
PMID 12634403
Abstract
This study shows a mechanism for the increase of signal transducer and activator of transcription 1 (STAT1) in Epstein-Barr virus-immortalized cells. Latent membrane protein 1 (LMP-1) expression was sufficient to induce STAT1 expression, DNA binding, and transcriptional activity. LMP-1-expressing cells can induce an increase in STAT1 expression in LMP-negative cells in the same culture, suggesting an indirect regulation of STAT1 expression. The increase in STAT1 expression is mediated by the C-terminal activating region 1 (CTAR-1) and/or CTAR-2 domains of LMP-1 and is inhibited by mutant IkappaB, demonstrating a role for NFkappaB in LMP-1-mediated STAT1 expression.
Related Materials