EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Effect of histamine on phasic and tonic contractions of isolated uterine tissue from pregnant women.
PMID 12634656
Abstract
To enhance our understanding of the uterotonic effect of histamine, we compared the effects of histamine on spontaneous phasic and tonic contractile activity of uterine strips from term pregnant nonlaboring women. Longitudinal uterine strips were used from the lower uterine segment of term pregnant nonlaboring women undergoing elective cesarean section. The concentration-response relationship to histamine (10(-8) to 10(-4) mol/L) was determined in spontaneously contracting strips or in strips contracted tonically with a protein kinase C activator (-)-indolactam V in the presence of H(1) receptor antagonist (S[+]-chlorpheniramine maleate), H(2) receptor antagonist (cimetidine), or solvent. Histamine increased spontaneous phasic myometrial contractions in a concentration-dependent manner. H(1), but not H(2), receptor antagonist significantly attenuated the response to histamine. Histamine significantly reduced tonic contractions of uterine strips induced by indolactam V. H(1) histamine receptor antagonist abolished relaxation, whereas H(2) histamine receptor antagonist had no effect. Histamine increases spontaneous, but inhibits tonic, contractions of uterine strips from term pregnant nonlaboring women. Both effects are mediated through activation of H(1) receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0661
(−)-Indolactam V, ≥96% (HPLC)
C17H23N3O2