EMAIL THIS PAGE TO A FRIEND
European journal of nuclear medicine and molecular imaging
Bilateral increase in striatal dopamine D2 receptor density in the 6-hydroxydopamine-lesioned rat: a serial in vivo investigation with small animal PET.
PMID 12634967
Abstract
Unilateral destruction of the substantia nigra by local application of 6-hydroxydopamine (6-OHDA) serves as an animal model for Parkinson's disease. In this study, the changes in neostriatal dopamine D(2) receptor density were investigated with a small animal positron emission tomograph (PET) before and after 6-OHDA lesion. PET scans were performed in 14 rats after injection of the D(2) receptor radioligand [(18)F] N-methylbenperidol. After the first scan (day 0), nigrostriatal pathways were lesioned by unilateral injections of 6-OHDA. Further PET scans were performed on days 2 and 14 post-lesion. For both striata, B(max) values were determined from saturation binding curves with non-linear regression analysis. In the striatum ipsilateral to the lesion, B(max) initially amounted to 19.3+/-1. 9 fmol/mg (mean+/-SD) and increased to 19.7+/-2.2 and 29.9+/-5.7 fmol/mg on days 2 and 14 post-lesion, respectively. Contralateral B(max) values increased from 19.2+/-2 fmol/mg prior to the lesion to 21.2+/-2.9 and 28.6+/-5.7 fmol/mg on days 2 and 14, respectively. On day 14, the ipsilateral saturation binding curve differed from the ipsilateral pre-lesion curve (P=0.04; F test). When the contralateral pre-lesion saturation binding curve was compared with the contralateral post-lesion curve on day 14, a P value of 0.08 was obtained. This first serial in vivo imaging study of 6-OHDA-lesioned rats showed a time-dependent increase in striatal D(2) receptor density on both sides, the increase being more pronounced ipsilateral to the lesion. This result implies that compensatory mechanisms in the intact hemisphere contribute to regenerative processes following nigrostriatal dopaminergic denervation. Overall, our findings show the feasibility of repetitive in vivo studies of striatal receptor density with a small animal tomograph. Moreover, the applied in vivo saturation binding technique provides a versatile method for the quantification of time-dependent changes in the concentration of receptor binding sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0470000 Benperidol, European Pharmacopoeia (EP) Reference Standard
C22H24FN3O2